missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System
Safe and approved storage of flammable hazardous substances in working areas.
Brand: asecos™ 3011600133715
2777.05 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
616 mm | |
524 x 1050 x 1742 mm | |
Wing | |
1968 mm | |
Door open arrest system | |
2 | |
Standard | |
451.50 kg | |
1050 mm | |
Steel |
3 x shelf, 1 x perforated insert(380 mm), 1 x 33 liter bottom collecting sump (sheet steel powder-coated) | |
524 mm | |
616 x 1196 x 1968 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm | |
S90.196.120.WDAS | |
3 | |
Powder-coated plain | |
1196 mm | |
Light Gray | |
Safety storage cabinet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction