missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Safety Can For Liquid Disposal

Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Safety Can For Liquid Disposal provides a better solution for the containment of liquid flammable or corrosive waste.

Brand:  Justrite™ 14265

 View more versions of this product

Product Code. 12769526

  • 304.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Built-in polyethylene fill gauge indicator pops up to warn you that the container is almost full
  • High-density polyethylene (HDPE) disposal cans provide superior chemical resistance and leak protection
  • Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig (0.2 and 0.35 bar)
  • Stainless-steel handle and hardware with latch-catch system hold the cap in the open position for easy filling and pouring
  • Large-mouth opening features a built-in stainless-steel flame arrester to guard against outside ignition
  • The broad-base design provides stability and resists accidental tip over
Specifications

Specifications

Safety Can
2 gal.
Liquid Disposal
Stainless steel handle and flame arrester; Fill gauge indicator.
Safety Can for Liquid Disposal
Red
HDPE
14.8 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.