Learn More
Justrite™ Safety Can For Liquid Disposal
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Safety Can For Liquid Disposal provides a better solution for the containment of liquid flammable or corrosive waste.
Brand:  Justrite™ 12772
 View more versions of this product
Product Code. 12923695
  • 602.00 € / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
High-density polyethylene (HDPE) disposal cans provide superior chemical resistance and leak protection. Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig (0.2 and 0.35 bar). Stainless steel handle and hardware with latch-catch system hold the cap in the open position for easy filling and pouring. Large-mouth opening features a built-in stainless steel flame arrester to guard against outside ignition. The broad-base design provides stability and resists accidental tip over.
Specifications
Specifications
Safety Can
5 gal.
Liquid Disposal
Stainless steel handle, faucet and flame arrester
Safety Can for Liquid Disposal
White
HDPE
20 in.