missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT HPLC Safety Caps

These safety caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
Brand: SCAT 107512
103.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of the connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every SafetyCap
Specifications
Autoclavable | |
Lab Bottles | |
PTFE | |
Safety Cap III |
HPLC Systems | |
3 PFA Fittings with 1.6/2.3/3.2 mm OD, 2 PFA Blind Plugs, 1 Air Valve | |
GL38 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction