missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ Safety Funnel 'ARNOLD' Replacement Lid
Replacement lid for the Safety Funnel 'ARNOLD' V2.0.
39.35 € - 49.65 €
Specifications
For Use With (Equipment) | ARNOLD Funnel |
---|---|
Includes | Replacement Lid |
Product Type | Funnel Lid |
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
17808733
|
SCAT™
319002 |
Black | HDPE-EC |
49.65 €
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
17888723
|
SCAT™
319001 |
Blue, White | HDPE |
39.35 €
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
Description
- Spare/replacement lid for use with the 'ARNOLD' V2.0 Safety Funnel
Specifications
ARNOLD Funnel | |
Replacement Lid | |
Funnel Lid |