Learn More
Deutsch Neumann™ Safety Gas Hoses
Provides safe and easy connection to gas burners. Deutsch Neumann™ Safety Gas Hoses consist of a rubber tube, threaded metal spiral and end sockets on both sides.
Brand:  Deutsch Neumann™ 3608050
 View more versions of this product
Product Code. 10077760
  • 41.65 € / 500mm
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Working pressure to 100 mbar
  • For gas burners meeting DIN 30 665
  • For all gases meeting DVGW Work Code G 260
Specifications
Specifications
Gas Hose
Gas burners meeting DIN 30 665 and all gases
500 mm
All Gases Meeting DVGW Work Code G 260