missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Safety Pipetting Ball
Buerkle™ Safety Pipetting Ball allows evacuation to take place via an automatic valve.
30.75 €
Specifications
Type | Ball |
---|---|
For Use With (Application) | Enables effective cleaning of the inside |
For Use With (Equipment) | up to 100 mL Pipettes |
Description
- Evacuation takes place via an automatic valve
- By pressing the valves, the liquid is drawn up, dispensed or purged
- The removable automatic valve also enables effective cleaning of the inside
- For all pipettes up to 100mL
- Available in various colours
Specifications
Ball | |
Enables effective cleaning of the inside | |
up to 100 mL Pipettes |