missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Safety Sign Hand Wash Instructions

An efficient solution with clear safety instructions to easily communicate COVID-19 countermeasures on work floors.

20.20 € - 21.40 €

Specifications

Material Laminated Polyester
Mounting Type Self-adhesive
Dimensions (L x W) 262 x 371 mm (W x H)
View More Specs

Products 10
Product Code Brand Languages Price Quantity & Availability  
Product Code Brand Languages Price Quantity & Availability  
16546414
View Documents
Brady™
306823
Dutch
21.40 €
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
16546444
View Documents
Brady™
306853
Norwegian This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16556414
View Documents
Brady™
306824
French
21.40 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
16556434
View Documents
Brady™
306844
Danish This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16566414
View Documents
Brady™
306825
German
21.40 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
16566434
View Documents
Brady™
306845
Finnish This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16576414
View Documents
Brady™
306826
English
21.10 €
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
 
16576434
View Documents
Brady™
306846
Swedish This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16586414
View Documents
Brady™
306827
Spanish This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16596414
View Documents
Brady™
306828
Italian
21.40 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
Description

Description

  • Provides clear, visual communication where it matters
  • Highlights the use of appropriate personal protection
  • Easily communicates social distancing and hygiene
Specifications

Specifications

Laminated Polyester
Self-adhesive
262 x 371 mm (W x H)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Safety Sign Hand Wash Instructions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.