Learn More
DWK Life Sciences Safety Standard Ground Socket, with thread
DURAN™ safety ground joints for increased safety requirements.
78.85 € - 296.00 €
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15806921
|
DWK Life Sciences
286592906 |
34 mm |
105.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15826921
|
DWK Life Sciences
286594504 |
50 mm |
296.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15876911
|
DWK Life Sciences
286591404 |
18 mm |
78.85 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15886911
|
DWK Life Sciences
286591901 |
22 mm |
79.55 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15896911
|
DWK Life Sciences
286592409 |
28 mm |
94.55 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
Description
Securing joints by using our KECK™-products is a well-known and proven method. In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN™ Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN™ Safety Joints offer a system to separate jammed cones and sockets. This minimizes the risk of breakage and related threats. The DURAN™ threaded sleeves also offer secure locking with DURAN™ standard cores.
- Threaded sleeve prevents unintentional loosening of the ground joint
- Firmly seated ground joints can be loosened more easily here. The danger of glass breakage and associated risks is thus minimized.
- Use in equipment for handling hazardous substances or working with pressure differences.g in the high vacuum range
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
10/Pk. |