Learn More
Sartorius Safety Weighing Cabinet without Filtration
Safety Weighing Cabinets ensure a safe working environment for laboratory staff without sacrificing ergonomics or precision in weighing. Personnel safety and reliable weighing results from a single source.
Brand:  Sartorius SWC900TNF
 View more versions of this product
Product Code. 16363086
  • 17285.00 € / Each
Explore more special offers
Description
Description
  • Appropriate protective measures must be taken to protect the health of all laboratory staff when they weigh hazardous compounds in powdered form.
  • The Sartorius SWC Safety Weighing Cabinet offers important advantages in design and construction over traditional laboratory hoods:
    • The SWC creates a contained area around the laboratory balance which prevents any air or finely powdered particulates from escaping into the operating personnel's work area.
    • At the same time, the constant inlet air velocity and the special cabinet construction both keep the air current practically turbulence-free, and, as a result, ensure consistent and repeatable weighing results.
    • The balance and weighing cabinet are perfectly matched to each other.
    • With its SWC Safety Weighing Cabinet, Sartorius has succeeded in uniting two otherwise contradictory requirements: maximum personnel protection and reliable weighing results.
    • Maximum personnel protection: Contained area around the laboratory balance which prevents any air or finely powdered particles from escaping into the operating personnel's work area
    • Reliable weighing results: Air current practically turbulence-free, and, consistent and repeatable weighing results.
    • Clear acrylic glass construction with perfect view of the balance and the sample to be weighed
    • Extremely stable base made of black granite absorbs vibrations
    • Contaminated materials can be disposed of safely in a double-bag system
    • Calibrated airflow alarm system with audit trail function sounds an alarm whenever the airflow velocity drops below a defined level
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Acrylic Glass
900 mm
29.5 in.
750 mm