Learn More
Sartorius™ Safety Weighing Cabinets
Ensure a safe working environment for laboratory staff without sacrificing ergonomics or precision in weighing. Sartorius™ Safety Weighing Cabinets offer important advantages in design and construction over traditional laboratory hoods.
Brand:  Sartorius™ SWC1200T
 View more versions of this product
Product Code. 10309444
  • 33375.00 € / Each
Explore more special offers
Description
Description
  • The balance and weighing cabinet are perfectly matched to each other
  • Maximum personnel protection: contained area around the laboratory balance which prevents any air or finely powdered particles from escaping into the operating personnel´s work area
  • Reliable weighing results: air current practically turbulence-free, and, consistent and repeatable weighing results.
  • Clear acrylic glass construction with perfect view of the balance and the sample to be weighed
  • Extremely stable base made of black granite absorbs vibrations
  • Contaminated materials can be disposed of safely in a double-bag system
  • Calibrated airflow alarm system with audit trail function sounds an alarm whenever the airflow velocity drops below a defined level
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Acrylic Glass
750 mm
Ensure a Safe Working Environment for Laboratory Staff Without Sacrificing Ergonomics or Precision in Weighing
1230 mm
Constant
.089 m3/sec. at 0.35 m/s face velocity with direction connection, .095 m3/sec. at 0.35 m/s face velocity with connection via fan filter
770 mm