missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ HPLC Safety Caps, V2.0
These HPLC Safety Caps allow you to extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
Brand: SCAT™ 307006
This item is not returnable.
View return policy
Description
SCAT™ Safety Caps V2.0 provide maximum safety for your HPLC solvent supply.
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every Safety Cap
Specifications
Autoclavable | |
HPLC Systems | |
2 x PFA 1.6/2.3/3.2 mm OD Fittings, PFA Blind Plug, Air Valve | |
GL 28 |
1.6 mm, 2.3 mm, 3.2 mm | |
Lab Bottle | |
PTFE | |
Safety Cap II |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction