missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HPLC Safety Caps, V2.0
GREENER_CHOICE

Product Code. 17848733 Shop All SCAT Europe Products
Click to view available options
Material:
PTFE
Polypropylene
Thread Size:
GL 28
GL 45
GL/S40
GL45
Includes:
1 x PFA 1.6/2.3/3.2 mm OD Fittings, 90° Angled Air Valve
1 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve
1 x PTFE 4 mm OD Fitting, Air Valve
2 x PFA 1.6/2.3/3.2 mm OD Fittings, 90° Angled Air Valve
2 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve
2 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve, PFA Blind Plug
2 x PP 1.6/2.3/3.3 mm OD Fittings, Air Valve, Change Indicator, PP Blind Plug, Labeling Field
3 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve
3 x PP 1.6/2.3/3.3 mm OD Fittings, Air Valve, Change Indicator, 2 x PP Blind Plug, Labeling Field
4 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve
4 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve, Change Indicator, 3 PFA Blind Plug, Labeling Field
6 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve
Unit Size:
1 sheet
Each
This item is not returnable. View return policy

Product Code. 17848733

Brand: SCAT™ 307410PP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These HPLC Safety Caps allow you to extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.

SCAT™ Safety Caps V2.0 provide maximum safety for your HPLC solvent supply.

  • High quality materials ensure maximum safety for lab and staff
  • 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
  • Stable construction for highest safety and easy handling
  • Chemically resistant against aggressive organic solvents
  • Attractive design and clear arrangement of connections
  • Improved design for easy connection of capillaries in various colors
  • Free air valve with change indicator included with every Safety Cap

Specifications

For Use With (Equipment) Lab Bottle
Includes 4 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve, Change Indicator, 3 PFA Blind Plug, Labeling Field
Material Polypropylene
Size (Metric) 1.6, 2.3, 3.2 mm O.D. Fittings
Thread Size GL 45
For Use With (Application) HPLC Systems
Type Safety Cap
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt