missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SafeVIEW MINI2 LED Transilluminator

Offers a safe way to view and document samples with a uniform blue light source. Fisherbrand™ SafeVIEW MINI2 LED Transilluminator will not cause damage to DNA or RNA that would normally be associated with UV light.

Brand:  Fisherbrand™ FSAFEVIEW-MINI2

Product Code. 15502506

  • 897.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Compact design
  • Even light source
  • Low cost
  • Lightweight at 1.52kg
Specifications

Specifications

470 nm LED Blue
200 x 200 x 28.7mm
154 x 154 mm
110/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.