missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SafTest™ Sample Consumables Platform

SafTest™ Sample Consumables Platform contains all consumables necessary to run alkenal, peroxide tests.

672.00 € - 1365.00 €

Specifications

Quantity 1 Ea.
Storage Requirements Room temperature
View More Specs

Includes: Printer paper, membranes, precision tissue wipers, solid glass beads, 15mL conical tubes, 10 to 100μL pipette tips, 200 to 1000μL pipette tips, 12mm culture tubes, 12mm culture tube caps, sharpie marke

Products 2
Product Code Brand For Use With (Application) Price Quantity & Availability  
Product Code Brand For Use With (Application) Price Quantity & Availability  
15792642
View Documents
MP Biomedicals™
07LC2000
Peroxide Testing, Alkenal Testing (Liquid Samples)
672.00 €
1 set
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
15712652
View Documents
MP Biomedicals™
07SC2000
Peroxide Testing, Alkenal Testing (Solid Samples)
1365.00 €
1 set
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
Description

Description

  • Consumable platform for conducting alkenal, peroxide testing
  • Contains all of consumables necessary to run solid, liquid samples
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Application area: Oil/fat degradation
Specifications

Specifications

1 Ea.
Room temperature
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.