Learn More
MP Biomedicals™ SafTest™ Test Kit
SafTest™ Test Kit measures percentage analytes in given samples. Choose from 6 different kits for use with SafTest instrument and consumable platforms and detection kits.
Brand: MP Biomedicals™ 07KTMA2000
Includes: Reagent A, Reagent B, Reagent C, Calibrators
Description
MP Biomedicals' Test Kits contain all reagents and calibrators necessary to run samples. A control is also included in each Test Kit. Each kit will allow for testing of approximately 100 samples. Sample dilution will likely be required for appropriate analysis.
- Choose from 6 different kits
- For use with SafTest instrument and consumable platforms
- Results in as little as 30 minutes
- Detection method: Bichromatic Photometer
- Test Kits contain all reagents and calibrators necessary to run samples.
- A control is also included in each Test Kit.
- Each kit will allow for testing of approximately 100 samples.
- Sample dilution will likely be required for appropriate analysis.
- Alkenal Test Kit
- Research Area: Lipid degradation and oxidation
- Contains all reagents, calibrators
- Free Fatty Acid Test Kit
- Research Area: Lipid degradation and oxidation
- Components: Reagent A, Reagent B, Calibrators, Control Analyte(s)
- Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
- Quantitation Range: 0.1 - 2.01% free fatty acid
- Malonaldehyde Test Kit
- Research Area: Lipid degradation and oxidation
- Components: Reagent A, Reagent B, Calibrators, Control
- Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
- Quantitation Range: 0.00 to 0.32 mg/kg of sample
- Percent Fat Test Kit
- Research Area: Lipid degradation and oxidation
- Components: Reagent A, Reagent B, Calibrators, Control Analytes(s)
- Material Required: SafTest Instrument Platform
- Quantitation Range: 0.06 to 0
- Peroxide Test Kit
- Research Area: Lipid degradation and oxidation
- Components: Reagent A, Reagent B, Reagent C, Calibrators, Control Analyte(s)
- Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
- Quantitation Range: 0.00 to 0.200 MEQ/kg of sample
- Total Glycerin Test Kit
- Research Area: Biodiesel
- Components: Reagent A, Reagent B, Calibrators, Control
- Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
- Quantitation Range: 0.008 to 0.06% of sample

Specifications
Malonaldehyde Testing | |
100 Tests | |
1 Ea. |
Bichromatic Photometer | |
Malonaldehyde | |
2°C to 8°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.