Learn More
Saint Gobain Life Sciences Chemware™ PTFE-Coated Forceps
Chemware™ PTFE-Coated Forceps for handling hot or chemically active materials.
Brand:  Saint Gobain Life Sciences D1069288
Product Code. 12361369
  • 28.80 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Nickel-plated steel covered with an extra heavy-duty coating of PTFE
  • Nonstick surface is inert
  • Coating prevents scratching glass and other delicate surfaces
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Straight
Chrome-plated Steel
Forceps
4.5 in.
114 mm
1 Ea.