missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Chemware™ PTFE Griffin Beakers

Product Code. 12356978
Click to view available options
Capacity (Metric):
1 mL
3 mL
5 mL
10 mL
30 mL
50 mL
100 mL
250 mL
400 mL
600 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemware™ PTFE Griffin Beakers are noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid.

  • Nonwettable, unbreakable and heat resistant
  • Easy to clean
  • Broad spout for easy pouring
  • Can be heated on hotplate up to 288°C (550°F) when proper care is exercised
  • Specially molded bottom allows rapid heat transfer
  • Unlike most PTFE beakers, these will boil water when covered

Applications:

Ideal for digestion

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 0.17 oz.
Capacity (Metric) 5 mL
Material PTFE
For Use With (Application) Ideal for Digestion
Height (English) 0.99 in.
Height (Metric) 2.53 cm
Includes Spout
Temperature (English) Max. Operating 550°F
Temperature (Metric) Max. Operating 288°C
Type Beaker
Style Griffin
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'