missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Chemware™ PTFE Laboratory Matting

Chemware™ PTFE Laboratory Matting is chemically inert—resists adhesion of practically any material.

Brand:  Saint Gobain Life Sciences D1069328

Product Code. 10686585

  • 291.00 € / 3m
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Corrosion- and break-resistant lining for benches, shelves, sinks, and drain boards helps to reduce glassware breakage
  • Useful as mesh bag for dipping parts in corrosive chemicals and solvents
  • Easy to clean
  • Diamond-shaped PTFE mesh sheeting is 0.06 in. thick (1.6 mm) with 0.25 in. (6 mm) openings
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lab Mat
10 ft.
12 in.
10 ft. x 12 in. (3 m x 310 mm)
0.062 in.
PTFE
3 m
310 mm
Corrosion and break resistant lining for benches, shelves, sinks, and drain boards helps to reduce glassware breakage
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.