missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 11944791
Click to view available options
Quantity:
2 g
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-02-8, 108-95-2
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O

Specifications

Melting Point -7.0°C
Color Yellow
Density 1.1600g/mL
Boiling Point 197.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. (Phenol) (GC)
Packaging Plastic drum
Linear Formula 2-(HO)C6H4CHO
Refractive Index 1.5720 to 1.5740
Quantity 5 kg
Beilstein 08,31
Merck Index 15,8462
Specific Gravity 1.16
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 122.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Salicylaldehyde
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.