Learn More
Salicylic acid, ACS, 99+%, Thermo Scientific Chemicals
31.80 € - 254.00 €
Chemical Identifiers
CAS | 69-72-7 |
---|---|
Molecular Formula | C7H6O3 |
Molecular Weight (g/mol) | 138.12 |
MDL Number | MFCD00002439 |
InChI Key | YGSDEFSMJLZEOE-UHFFFAOYSA-N |
Synonym | salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone |
PubChem CID | 338 |
ChEBI | CHEBI:16914 |
IUPAC Name | 2-hydroxybenzoic acid |
SMILES | OC(=O)C1=CC=CC=C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307358
|
Thermo Scientific Alfa Aesar
030782.22 |
100 g |
31.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397348
|
Thermo Scientific Alfa Aesar
030782.36 |
500 g |
85.50 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15411177
|
Thermo Scientific Alfa Aesar
030782.0E |
2500 g |
254.00 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Salicylic acid is used as a food preservative as well as used in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in the dye industry for making azo direct dyes and acidic mordant dyeing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69-72-7 | |
138.12 | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
338 | |
2-hydroxybenzoic acid |
C7H6O3 | |
MFCD00002439 | |
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
CHEBI:16914 | |
OC(=O)C1=CC=CC=C1O |
Specifications
69-72-7 | |
211°C (20mmHg) | |
MFCD00002439 | |
14,8332 | |
Soluble in carbon terachloride,benzene,propanol,absolute ethanol,acetone,toluene and ether; Slightly soluble in cold water; More soluble in boiling water; Water solubility increased by Na3PO4,alkali acetates or alkali citrates. | |
OC(=O)C1=CC=CC=C1O | |
138.12 | |
CHEBI:16914 | |
≥99% | |
1.443 g/mL | |
Powder |
158°C to 160°C | |
C7H6O3 | |
774890 | |
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
2-hydroxybenzoic acid | |
338 | |
138.12 | |
ACS | |
157°C (315°F) | |
Salicylic acid |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P201-P202-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P501c
H302-H318-H361d
missing translation for 'einecsNumber' : 200-712-3
missing translation for 'rtecsNumber' : VO0525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only