missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylic Acid, For analysis ACS, +99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifica
Specifica
Melting Point | 158.0°C to 161.0°C |
Boiling Point | 211.0°C (20.0 mmHg) |
Color | White |
Assay Percent Range | 99+% |
Beilstein | 10, 43 |
Merck Index | 15, 8469 |
Formula Weight | 138.12 |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Quantity | 500 g |
Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- Salicylic acid
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'