missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylic acid, Honeywell Fluka™
66.02 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
1KG POLY BOTTLE
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
CAS | 69-72-7 |
Appearance | Complying |
PubChem CID | 338 |
ChEBI | CHEBI:16914 |
Percent Purity | 99.5 to 100.5 % |
Grade | Reagent Pharmacopeia Acids |
Quantity | 1 kg |
Loss on Drying | Max. 0.3 % |
Packaging | Plastic Bottle |
Physical Form | Crystalline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'