missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylic acid, sodium salt, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Acidity | 2mL 0.01N NaOH max. |
Melting Point | 200.0°C |
Color | Brown to Brown-Red |
Loss on Drying | 0.5% max. (1g, 105°C) |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Plastic bottle |
Linear Formula | 2-(HO)C6H4CO2Na |
Quantity | 50 g |
Beilstein | 10, 43 |
Show More |
Safety and Handling
Product Identifier
- Salicylic acid, sodium salt
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction