missing translation for 'onlineSavingsMsg'
Learn More

Cole Parmer Salt Brine Hydrometer

Salt Brine Hydrometer is equivalent to H-B Instrument 618090000

Brand:  Cole Parmer 08297-38

Product Code. 15355975

  • 39.35 € / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Salt Brine Hydrometer is equivalent to H-B Instrument 618090000

  • Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST.
  • Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
  • Temperature of Standardization 60°F/60°F
  • 0/100 Percent by Saturation
  • Metal Ballast
  • Individually Serialized
Specifications

Specifications

Hydrometer
280 mm
Specific Gravity Measurement
0.01
Metal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole Parmer Salt Brine Hydrometer > hydrometer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.