Learn More
Samarium, 99.9%, (trace metal basis), 20 to 40 mesh, Thermo Scientific Chemicals
160.00 € - 505.00 €
Chemical Identifiers
CAS | 7440-19-9 |
---|---|
Molecular Formula | Sm |
Molecular Weight (g/mol) | 150.36 |
MDL Number | MFCD00011233 MFCD00151299 |
InChI Key | KZUNJOHGWZRPMI-UHFFFAOYSA-N |
Synonym | samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l |
PubChem CID | 23951 |
ChEBI | CHEBI:33374 |
IUPAC Name | samarium |
SMILES | [Sm] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10450721
|
Thermo Scientific Acros
294780100 |
10 g | Plastic bottle |
160.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10115553
|
Thermo Scientific Acros
294780500 |
50 g | Plastic bottle |
505.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7440-19-9 | |
150.36 | |
KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
23951 | |
samarium |
Sm | |
MFCD00011233 MFCD00151299 | |
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l | |
CHEBI:33374 | |
[Sm] |
Specifications
1778.0°C | |
7440-19-9 | |
Powder | |
Sm | |
MFCD00011233 MFCD00151299 | |
17,305 | |
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l | |
KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
samarium | |
23951 | |
150.36 | |
Trace Metal Basis |
1072.0°C | |
Gray | |
99.9% | |
Sm | |
3132 | |
15, 8484 | |
Solubility in water: reacts. Other solubilities: forming hydrogen | |
[Sm] | |
150.36 | |
CHEBI:33374 | |
99.9% | |
Samarium, 40 Mesh |
Safety and Handling
GHS H Statement
In contact with water releases flammable gas.
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a dry place.
Store in a closed container.
In case of fire: Use dry sand,dry chemical or alcohol-resistant foam for extinction.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-128-7
- Samarium, powder
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
- Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
Hazard Statement
- H228-Flammable solid.
- H261-In contact with water releases flammable gases.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only