Learn More
Samarium(II) iodide, 0.1M solution in THF, stabilized with Samarium powder, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 294991000
Code : 29
Additional Details : CAS Number : 32248-43-4 Weight : 0.09220kg Transport : UN number : 2056 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Samarium(II) iodide, 0.1M solution in THF | |
Blue-Green | |
Solution | |
I2Sm | |
MFCD00058873 | |
15, 8486 | |
404.17 | |
AcroSeal™ Glass Bottle | |
0.9220g/mL |
0.1M solution in THF, Stabilized with samarium powder | |
32248-43-4,109-99-9 | |
100 mL | |
SmI2 | |
−17°C | |
404.17 | |
0.1M, exact strength on the certificate of analysis | |
0.922 |
Safety and Handling
- Samarium(II) iodide, 0.1M solution in THF, stabilized with Samarium powder
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only