missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Chemical Identifiers

CAS 13517-12-9
Molecular Formula Br3H12O6Sm
Molecular Weight (g/mol) 498.162
MDL Number MFCD00192430
InChI Key LXJDCZVUWJNXRL-UHFFFAOYSA-K
Synonym acmc-20ajrk, samarium bromide hexahydrate, samarium 3+ bromide-water 1/3/6, samarium iii bromide hexahydrate, reacton™
PubChem CID 57348178
IUPAC Name samarium(3+);tribromide;hexahydrate
SMILES O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11362558
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014603.14
25 g N/A
N/A
11372558
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014603.22
100 g N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13517-12-9
498.162
LXJDCZVUWJNXRL-UHFFFAOYSA-K
57348178
O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]
Br3H12O6Sm
MFCD00192430
acmc-20ajrk, samarium bromide hexahydrate, samarium 3+ bromide-water 1/3/6, samarium iii bromide hexahydrate, reacton™
samarium(3+);tribromide;hexahydrate

Specifications

>640°C
Crystalline
MFCD00192430
LXJDCZVUWJNXRL-UHFFFAOYSA-K
samarium(3+);tribromide;hexahydrate
57348178
99.9%
Hygroscopic
Samarium(III) bromide hexahydrate
13517-12-9
Br3H12O6Sm
acmc-20ajrk, samarium bromide hexahydrate, samarium 3+ bromide-water 1/3/6, samarium iii bromide hexahydrate, reacton™
O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]
498.162
498.15 (390.06 Anhydrous)
Reacton™
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 237-347-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only