Learn More
Samarium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
63.90 € - 620.00 €
Chemical Identifiers
CAS | 10361-82-7 |
---|---|
Molecular Formula | Cl3Sm |
Molecular Weight (g/mol) | 256.71 |
MDL Number | MFCD00011235 |
InChI Key | BHXBZLPMVFUQBQ-UHFFFAOYSA-K |
IUPAC Name | samarium(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Sm+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338707
|
Thermo Scientific Alfa Aesar
035804.04 |
2 g |
63.90 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328707
|
Thermo Scientific Alfa Aesar
035804.09 |
10 g |
180.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11318707
|
Thermo Scientific Alfa Aesar
035804.18 |
50 g |
620.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This compound named Samarium(III) chloride has few practical applications but is used in laboratories for research on new compounds of samarium. Samarium(III) chloride is used for the preparation of samarium metal, which has a variety of uses, notably in magnets. Anhydrous SmCl3 is mixed with sodium chloride or calcium chloride to give a low melting point eutectic mixture. Electrolysis of this molten salt solution gives the free metal. Samarium(III) chloride can also be used as a starting point for the preparation of other samarium salts. The anhydrous chloride is used to prepare organometallic compounds of samarium, such as bis(pentamethylcyclopentadienyl)alkylsamarium(III) complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
10361-82-7 | |
256.71 | |
BHXBZLPMVFUQBQ-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[Sm+3] |
Cl3Sm | |
MFCD00011235 | |
samarium(3+) trichloride |
Specifications
(decomposition) | |
10361-82-7 | |
Cl3Sm | |
14,8347 | |
BHXBZLPMVFUQBQ-UHFFFAOYSA-K | |
samarium(3+) trichloride | |
256.71 | |
Ampule | |
4.46 g/mL at 16°C | |
Samarium(III) chloride, Ultra dry |
678°C | |
Powder | |
MFCD00011235 | |
It is soluble in water. | |
[Cl-].[Cl-].[Cl-].[Sm+3] | |
256.71 | |
99.9% | |
Hygroscopic | |
(REO) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 233-797-0
missing translation for 'rtecsNumber' : VP2625000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only