missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11367637
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in NMR spectroscopy for spectra resolution and is used for doping purpose. It is also used as a chemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13759-83-6
Molecular Formula H12N3O15Sm
Molecular Weight (g/mol) 444.46
MDL Number MFCD00149857
InChI Key HDCOFJGRHQAIPE-UHFFFAOYSA-N
Synonym samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
PubChem CID 203081
SMILES O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 78°C to 79°C
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water and acetonitrile.
IUPAC Name samarium(3+);trinitrate;hexahydrate
Formula Weight 444.45 (336.36 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Yellow crystals
Density 2.375 g/cm3 at 20°C
Chemical Name or Material Samarium(III) nitrate hexahydrate
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Samarium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.