missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

100.50 € - 268.34 €

Chemical Identifiers

CAS 13759-83-6
Molecular Formula H12N3O15Sm
Molecular Weight (g/mol) 444.46
MDL Number MFCD00149857
InChI Key HDCOFJGRHQAIPE-UHFFFAOYSA-N
Synonym samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
PubChem CID 203081
SMILES O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377637
SDS View Documents
Thermo Scientific Alfa Aesar
012906.14
25 g
100.50 €
25g
Please to purchase this item. Need a web account? Register with us today!
11367637
SDS View Documents
Thermo Scientific Alfa Aesar
012906.22
100 g
268.34 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in NMR spectroscopy for spectra resolution and is used for doping purpose. It is also used as a chemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13759-83-6
444.46
HDCOFJGRHQAIPE-UHFFFAOYSA-N
203081
H12N3O15Sm
MFCD00149857
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

78°C to 79°C
Crystalline
H12N3O15Sm
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
HDCOFJGRHQAIPE-UHFFFAOYSA-N
samarium(3+) hexahydrate trinitrate
203081
Odorless
Yellow crystals
Samarium(III) nitrate hexahydrate
13759-83-6
99.9% (REO)
MFCD00149857
Soluble in water and acetonitrile.
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
444.46
444.45 (336.36 Anhydrous)
Hygroscopic
2.375
Videos
Safety and Handling

Safety and Handling

P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P380+P375-P372-P380-P501c

H272-H315-H320-H335

missing translation for 'dotInformation' : DOT Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 233-798-6

missing translation for 'rtecsNumber' : VP3150000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.