missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11396268
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Samarium(III) nitrate hydrate is used as a catalyst in von Pechmann condensation for the preparation of coumarins. Further, it finds use in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. It is also used as a laboratory reagent. In addition to this, it is used in NMR spectroscopy for spectral resolution and is used for doping purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13759-83-6
Molecular Formula H12N3O15Sm
Molecular Weight (g/mol) 444.46
MDL Number MFCD00149857
InChI Key HDCOFJGRHQAIPE-UHFFFAOYSA-N
Synonym samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
PubChem CID 203081
SMILES O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 78°C to 79°C
Color Yellow
pH ∼3
Physical Form Crystalline Aggregates
Quantity 25 g
UN Number UN1477
Solubility Information Soluble in water and acetonitrile.
IUPAC Name samarium(3+);trinitrate;hexahydrate
Formula Weight 336.36 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 2.375 g/mL
Assay (REO)
Chemical Name or Material Samarium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.