missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) oxalate decahydrate, 99.9% (REO), Thermo Scientific™

51.00 € - 158.00 €

Chemical Identifiers

CAS 14175-03-2
Molecular Formula C6H26O22Sm2
Molecular Weight (g/mol) 750.97
MDL Number MFCD00150489
InChI Key IKPDQYODFRQYAD-UHFFFAOYSA-N
Synonym samarium iii oxalate hydrate 2:3:10, ethanedioic acid,samarium 3+ salt, hydrate 3:2:10
PubChem CID 121488066
IUPAC Name oxalic acid;samarium;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Sm].[Sm].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342558
SDS View Documents
Thermo Scientific Alfa Aesar
014602.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11352558
SDS View Documents
Thermo Scientific Alfa Aesar
014602.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14175-03-2
750.97
IKPDQYODFRQYAD-UHFFFAOYSA-N
121488066
O.O.O.O.O.O.O.O.O.O.[Sm].[Sm].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
C6H26O22Sm2
MFCD00150489
samarium iii oxalate hydrate 2:3:10, ethanedioic acid,samarium 3+ salt, hydrate 3:2:10
oxalic acid;samarium;decahydrate

Specifications

14175-03-2
Crystalline
Sm2(C2O4)3·10H2O
UN3288
IKPDQYODFRQYAD-UHFFFAOYSA-N
oxalic acid;samarium;decahydrate
121488066
99.9%
Samarium(III) oxalate decahydrate
White
C6H26O22Sm2
MFCD00150489
samarium iii oxalate hydrate 2:3:10, ethanedioic acid,samarium 3+ salt, hydrate 3:2:10
O.O.O.O.O.O.O.O.O.O.[Sm].[Sm].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
750.97
744.93 (564.78 Anhydrous)
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.