Learn More
Samarium(III) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Description
The biocompatibility of natural samarium (III) oxide, which has previously been used for treatment in bone-related diseases was determined as a first step in its evaluation as a bone implant material. The rates and products of the purely heterogeneous oxidations of C2H6 (g) and C2H4 (g) on Sm2O3 in the presence of O2 (g) were investigated in a very low-pressure flow reactor by on-line molecular beam mass spectrometry, about 1000±100 K. Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. Also, it is used as a neutron absorber in control rods for nuclear power reactors. The oxide catalyzes dehydration of acyclic primary alcohols to aldehydes and ketones. Another use involves preparation of other samarium salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2300°C |
Color | White |
Physical Form | Sintered Lump |
Quantity | 50 g |
Assay Percent Range | 99.95% (metals basis) |
Solubility Information | Insoluble in water |
Formula Weight | 348.7 |
Refractive Index | 1.97 |
Appearance | Off-white 3-12mm Sintered Lump |
Chemical Name or Material | Samarium(III) oxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.