missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) phosphate hydrate, 99.9% (REO), Thermo Scientific™

Chemical Identifiers

CAS 14913-18-9
MDL Number MFCD05665148
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314997
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041992.04
2 g N/A
N/A
11324997
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041992.09
10 g N/A
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14913-18-9
MFCD05665148

Specifications

14913-18-9
MFCD05665148
99.9%
Samarium(III) phosphate hydrate
Powder
246.41 (Anhydrous)
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 236-698-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications