missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) sulfate octahydrate, REacton™, 99.9% (REO)

Product Code. p-7049527
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11332558

Brand: Thermo Scientific Alfa Aesar 014601.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13465-58-2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-58-2
MDL Number MFCD00011236

Specifications

Melting Point 450°C (in -8H2O)
Physical Form Crystalline
Quantity 25 g
Merck Index 14,8347
Solubility Information Sparingly soluble in water
Formula Weight 733.00 (588.88 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Samarium(III) sulfate octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.