missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Sambucus Nigra Lectin [Cy3]

Product Code. 18316547 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18316547

Brand: Novus Biologicals™ CL1303NB

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Concentration 1 mg/mL
For Use With (Application) Immunocytochemistry/Immunofluorescence
Formulation 10 mM HEPES (pH 7.5), 0.15 M NaCl, 0.1 mM CaCl2, proprietary stabilizer
Quantity 1 mg
Storage Requirements Store at 4°C in the dark.
Inhibitor Properties A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37C and centrifuge before use.
Research Category Glycobiology
Protein Sambucus Nigra Lectin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis