Learn More
Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes
GREENER_CHOICE
Perform precise dispensing of volumes from 20 to 400 μL by using these exact volume transfer pipettes. Made of shatterproof, nontoxic polyethylene.
Brand:  Thermo Scientific™ 191NL
34.44 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
 View more versions of this product
Product Code. 15397803
  • 38.30 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Perform precise dispensing of volumes from 20 to 400 μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
  • Shatterproof, nontoxic polyethylene
  • Nonsterile
  • Low affinity surface
  • Freezer ready
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Samco Exact Volume Transfer Pipettes; 250μL Exact Volume Transfer Pipettes, Non-sterile (500/Pack, 5000/Case)
Polyethylene
250 μL
3 mm
Non-sterile
10.19 cm