missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Sample Collecting Bag

For use with disposable Sack Samplers to collect samples taken through the side of a sack.

Brand:  Sampling Systems™ 8080A-450

Product Code. 15611187

  • 312.00 € / Pack of 1000
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 450mL capacity
  • Accessory item
  • Sterile, for hygienic use
  • Provided 1000 bags/pack
Specifications

Specifications

450 mL
Sterile
Sack Samplers
1000/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers