Learn More
Velp Scientifica™ Sample Racks for Digestion Units
Facilitates nitrogen analysis and protein determination. Velp Scientifica™ Sample Racks for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
Brand:  Velp Scientifica™ A00000168
 View more versions of this product
Product Code. 11795654
  • 297.00 € / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available with or without heat shields
  • Available to fit a number of tube sizes and counts
Specifications
Specifications
Sample Rack
DK20 and DKL 20 Digesters