Learn More
Buerkle™ Sampler Mole with Handle and Bar
Very robust model with specially hardened forged drill bits. For determining soil properties, moisture and structure. The shape of the Mole's drill bits ensures minimum friction when drilling and removing the Mole from the ground. This reduces physical effort to a minimum.
Brand: Buerkle™ 5350-1005
316.09 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Alert:
Please order drill bits separately!
Description
The shape of the Mole's drill bit ensures minimum friction when drilling and removing the Mole from the ground. Seven different drill bits are available, each of which is designed for a particular soil type.- Soil sampler
- Seven different drill bits available
Specifications
75 cm |