missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Sampling Dippers
PTFE construction with reinforced rim and spout. PTFE handle includes steel core.
Brand: BRAND™ 90438
149.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- PTFE construction
- PTFE handle includes steel core
- Includes spout and reinforced rim
Specifications
Dip Sampler | |
With reinforced rim and spout | |
55 mm |
PTFE | |
100 mL | |
65 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction