missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ V4A Stainless Steel Inserts for Novartos Uno

Buerkle™ V4A Stainless Steel Inserts for Novartos Uno permit the withdrawal of a double sample without having to insert the lance repeatedly into the bulk material.

Brand:  Buerkle™ 5358-0140

148.98 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15336287

  • 175.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel AISI 316L
  • Fit precisely
  • Can easily be replaced
  • Available in volumes from 0 to 4.0mL
Specifications

Specifications

Insert
Autoclavable
4 mL
FDA
Novartos Uno
Stainless Steel (V4A)
Equipment for Novartos Uno sampler
Silver
Withdrawling Samples
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.