missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ SAN Collection Pitchers

Styrene Acrylonitrile Resin (SAN) construction. Vitlab™ SAN Beakers include easy-grip handle and white PC lid.

Brand:  Vitlab™ 97891

Product Code. 15223059

  • 105.00 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Raised scale allows for easy assessment of liquid
  • Includes stable, easy-grip handle and white PC lid.
  • 2000mL capacity
  • Diameter: 150mm
  • Height: 220mm
  • SAN construction
Specifications

Specifications

Yes
Clear
20 mL
White PC lid
2000 mL
SAN
150 mm
220 mm
6/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers