missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, for analysis, 40-100 mesh, Thermo Scientific Chemicals
36.58 € - 375.87 €
Chemical Identifiers
CAS | 14808-60-7 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10619014
|
Thermo Scientific Acros
393302500 |
250 g | Plastic bottle |
36.58 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10340494
|
Thermo Scientific Acros
393300010 |
1 kg | Plastic bottle |
93.06 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13468538
|
Thermo Scientific Acros
393300050 |
5 kg | Plastic bottle |
375.87 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidChemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
2230.0°C | |
14808-60-7 | |
Crystals | |
SiO2 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08 | |
Sand |
1610.0°C | |
Beige to Gray | |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
Solubility in water: insoluble | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Analytical |
Safety and Handling
missing translation for 'einecsNumber' : 238-878-4
RUO – Research Use Only