missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh, Fisher Chemical™
23.80 € - 66.80 €
Chemical Identifiers
CAS | 14808-60-7 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10355482
|
Fisher Chemical
S/0365/60 |
1 kg |
23.80 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10020262
|
Fisher Chemical
S/0365/63 |
3 kg |
66.80 €
3kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh
Chemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
2230°C | |
14808-60-7 | |
Solid | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08g/mol |
1610°C | |
Beige | |
O2Si | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Plastic powder jar |