Learn More
Notrax Sani-Flex™ Specially Designed for the Lab Industry
Specially designed for the lab industry, Sani-Flex™ anti-fatigue matting provides the optimum combination of comfort, anti-slip surface, quality and durability, as well as oil/grease and chemical resistance. Its smooth surface and rounded off edges make it easy to clean.
87.98 € - 152.00 €
Specifications
Color Gray
Material Nitrile Rubber
Type Industrial and Commercial Matting
View More Specs
Products 2
Product Code Brand Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Price Quantity & Availability  
17792178
View Documents
Notrax
526S1824GY
61 cm 46 cm
87.98 €
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
17782178
View Documents
Notrax
526S0023GY
91 cm 61 cm
152.00 €
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
Description
Description
  • Specially designed for the lab industry, Sani-Flex™ anti-fatigue matting provides the optimum combination of comfort, anti-slip surface, quality and durability, as well as oil/grease and chemical resistance
  • Its smooth surface and rounded off edges make it easy to clean
  • Made of tough rubber compounds, designed to yield a long service life
  • Aggressive slip-resistant surface
Specifications
Specifications
Gray
Nitrile Rubber
Industrial and Commercial Matting
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Notrax Sani-Flex™ Specially Designed for the Lab Industry