missing translation for 'onlineSavingsMsg'
Learn More

Notrax Sani-Trax™ Disinfecting Foot Bath Entrance Mat, Black

Notrax Sani-Trax™ Disinfecting Foot Bath Entrance Mat cleans and sanitizes shoes, recommended at entrances to all food processing areas. As traffic passes, rubber tips bend to immerse soles in disinfectant solution, a double action that keeps contamination from reaching food processing areas.

Brand:  Notrax 346S1824BL

 View more versions of this product

Product Code. 17782018

  • 46.00 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • This mat cleans and sanitizes shoes, recommended at entrances to all food processing areas
  • Thousands of flexible fingers provide an automatic scrubbing action that dislodges contaminants from footwear
  • As traffic passes, rubber tips bend under pressure to immerse soles in disinfectant solution, a double action that keeps contamination from reaching food processing areas
  • Convenient mat size fits most doorways
  • Suction cups on underside prevents mat from shifting
Specifications

Specifications

Black
24 cm
Industrial and Commercial Matting
9.4 in.
Natural Rubber
18 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.