missing translation for 'onlineSavingsMsg'
Learn More
SANT-1, Tocris Bioscience™ Product Code.: 18085972

SANT-1, Tocris Bioscience™

Product Code. 18085972
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18085972

Brand: Tocris Bioscience 1974/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of hedgehog (Hh) signaling; antagonizes smoothened activity

SANT-1 is a potent, cell-permeable inhibitor of Sonic hedgehog (Shh) signaling; high affinity antagonist of smoothened activity (KD = 1.2 nM). Inhibits smoothened agonist effects with an IC50 of 20 nM (in Shh-LIGHT2 cells). Has been used in protocols to generate pancreatic β cells from hPSCs.

For more information about how SANT-1 may be used, see our protocol:

Generation of β cells from hPSCs
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304909-07-7
Molecular Formula C23H27N5
Molecular Weight (g/mol) 373.50
MDL Number MFCD01827306
InChI Key FOORCIAZMIWALX-UHFFFAOYSA-N
Synonym sant-1, shh signaling antagonist v, 4-benzyl-piperazin-1-yl-3,5-dimethyl-1-phenyl-1h-pyrazol-4-ylmethylene-amine, e-4-benzyl-n-3,5-dimethyl-1-phenyl-1h-pyrazol-4-yl methylene piperazin-1-amine, n-3,5-dimethyl-1-phenyl-1h-pyrazol-4-yl methylene-4-phenylmethyl-1-piperazinamine, n-4-benzylpiperazin-1-yl-1-3,5-dimethyl-1-phenyl-1h-pyrazol-4-yl methanimine, e-n-4-benzylpiperazin-1-yl-1-3,5-dimethyl-1-phenylpyrazol-4-yl methanimine, 4-benzyl-n-3,5-dimethyl-1-phenyl-1h-pyrazol-4-yl methylene piperazin-1-amine, 4-benzyl-n-e-3,5-dimethyl-1-phenyl-1h-pyrazol-4-yl methylidene piperazin-1-amineShow MoreShow Less
PubChem CID 6878030
IUPAC Name N-(4-benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanimine
SMILES CC1=NN(C(C)=C1C=NN1CCN(CC2=CC=CC=C2)CC1)C1=CC=CC=C1

Specifications

Quantity 50 mg
Formula Weight 373.5
Percent Purity >99%
Chemical Name or Material SANT-1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SANT-1, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.