Learn More
Gilson™ Pharmed™ Pump Tubing
For use in the pharmaceutical and food industries
Brand: Gilson™ F1825101
This item is not returnable.
View return policy
Description
- Polypropylene
Specificatie
Pump Tubing | |
Santoprene | |
For MF1 and MF4 Pump Heads | |
2.5mL/min. |
0.5 mm | |
40 cm | |
4 Pack |